1,1,1,5,5,5-Hexafluoroacetylacetone 98%
Supplier: Apollo Scientific
Danger
Synonyms: Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA
PC4590-100G PC4590-25G
APOSPC4590-100GEA 281 EUR
APOSPC4590-100G APOSPC4590-25G
1,1,1,5,5,5-Hexafluoroacetylacetone 98%
1,1,1,5,5,5-Hexafluoroacetylacetone
Formula: C₅H₂F₆O₂
MW: 208.06 g/mol
Boiling Pt: 70 °C
Density: 1.486 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00000426
CAS Number: 1522-22-1
EINECS: 216-191-0
UN: 2920
ADR: 8+3 II