2-Methoxy-4-(trifluoromethyl)benzoic acid 98%
Supplier: Apollo Scientific
Warning
PC4602-5G PC4602-1G
APOSPC4602-5GEA 273 EUR
APOSPC4602-5G APOSPC4602-1G
2-Methoxy-4-(trifluoromethyl)benzoic acid 98%
2-Methoxy-4-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD04115976
CAS Number: 448-36-2