4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether

Supplier: Apollo Scientific
Warning

PC4618-1G PC4618-5G
APOSPC4618-1GEA 201 EUR
APOSPC4618-1G APOSPC4618-5G
4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether
4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether
Formula: C₁₄H₉F₃N₂S
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00793554
CAS Number: 320421-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR