1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride
Supplier: Apollo Scientific
Warning
Synonyms: O-(4-(Trifluoromethyl)benzyl)hydroxylamine hydrochloride
PC4632-1G PC4632-5G
APOSPC4632-1GEA 294 EUR
APOSPC4632-1G APOSPC4632-5G
1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride
1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride
Formula: C₈H₉ClF₃NO
MW: 227.61 g/mol
Melting Pt: 325…328 °C
Storage Temperature: Ambient
MDL Number: MFCD00231881
CAS Number: 321574-29-2