ω-Bromo-2-fluoroacetophenone 98%
Supplier: Apollo Scientific
Danger
Synonyms: Bromomethyl o-fluorophenyl ketone, 2-Bromo-2'-fluoroacetophenone, α-Bromo-2-fluoroacetophenone, 2-Fluorophenacyl bromide, 2-Bromo-1-(2-fluorophenyl)ethan-1-one, 2-Bromo-1-(2-fluorophenyl)ethanone
PC4747-5G PC4747-25G
APOSPC4747-5GEA 31.9 EUR
APOSPC4747-5G APOSPC4747-25G
ω-Bromo-2-fluoroacetophenone 98%
ω-Bromo-2-fluoroacetophenone
Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 83…85 °C
Melting Pt: 25…30 °C
Density: 1.574 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00278796
CAS Number: 655-15-2
UN: 3261
ADR: 8 III