α-Bromo-2-(trifluoromethoxy)toluene 97%
Supplier: Apollo Scientific
Danger
PC4829-100G PC4829-25G PC4829-5G
APOSPC4829-100GEA 202 EUR
APOSPC4829-100G APOSPC4829-25G APOSPC4829-5G
α-Bromo-2-(trifluoromethoxy)toluene 97%
α-Bromo-2-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Boiling Pt: 222
Melting Pt: 32
Density: 1.598 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00236321
CAS Number: 198649-68-2
UN: 3265
ADR: 8 III