3-Trifluoromethoxy-N-(trimethylsiliyl)aniline
Supplier: Apollo Scientific
PC48314-1G PC48314-2.5G PC48314-5G PC48314-10G
APOSPC48314-1GEA 236 EUR
APOSPC48314-1G APOSPC48314-2.5G APOSPC48314-5G APOSPC48314-10G
3-Trifluoromethoxy-N-(trimethylsiliyl)aniline
3-Trifluoromethoxy-N-(trimethylsiliyl)aniline
Formula: C₁₀H₁₄F₃NOSi
MW: 249.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14666453
CAS Number: 561304-47-0