1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-methylbenzene
Supplier: Apollo Scientific
Danger
Synonyms: 4-Methyl-2,3,5,6-tetrafluorobenzyl bromide
PC4837-1G PC4837-25G PC4837-5G
APOSPC4837-1GEA 183 EUR
APOSPC4837-1G APOSPC4837-25G APOSPC4837-5G
1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-methylbenzene
1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-methylbenzene
Formula: C₈H₅BrF₄
Boiling Pt: 49 °C/0.4mmHg
Melting Pt: 38…43 °C
Flash Pt: 92.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03001155
CAS Number: 92814-00-1
UN: 3261
ADR: 8,II