4-Bromo-2-chloro-1-(trifluoromethoxy)benzene 97%
Supplier: Apollo Scientific
Warning
PC48424-10G PC48424-1G PC48424-5G
APOSPC48424-10GEA 39.3 EUR
APOSPC48424-10G APOSPC48424-1G APOSPC48424-5G
4-Bromo-2-chloro-1-(trifluoromethoxy)benzene 97%
4-Bromo-2-chloro-1-(trifluoromethoxy)benzene
Formula: C₇H₃BrClF₃O
MW: 275.4503 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699114
CAS Number: 158579-80-7