1-Bromomethyl-2,2-difluorocyclopropane
Supplier: Apollo Scientific
Warning
PC49013-1G PC49013-5G
APOSPC49013-1GEA 350 EUR
APOSPC49013-1G APOSPC49013-5G
1-Bromomethyl-2,2-difluorocyclopropane
1-Bromomethyl-2,2-difluorocyclopropane
Formula: C₄H₅BrF₂
MW: 170.985 g/mol
Boiling Pt: 108…109 °C
Storage Temperature: Ambient
MDL Number: MFCD06797749
CAS Number: 77613-65-1
UN: 1992
ADR: 3+6.1 III