2-(Difluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Warning
PC49143-10G PC49143-5G
APOSPC49143-10GEA 366 EUR
APOSPC49143-10G APOSPC49143-5G
2-(Difluoromethyl)pyridine 97%
2-(Difluoromethyl)pyridine
Formula: C₆H₅F₂N
Density: 1.204 g/cm³ (25 °C)
Flash Pt: 41.7 °C
Storage Temperature: Ambient
MDL Number: MFCD11226632
CAS Number: 114468-01-8
UN: 1993
ADR: 3,III