Ethyl 2-fluoroisobutyrate
Supplier: Apollo Scientific
Danger
Synonyms: Ethyl 2-fluoro-2-methylpropanoate
PC49233-25G PC49233-5G PC49233-1G
APOSPC49233-25GEA 0 EUR
APOSPC49233-25G APOSPC49233-5G APOSPC49233-1G
Ethyl 2-fluoroisobutyrate
Ethyl-2-fluoroisobutyrate
Formula: C₆H₁₁FO₂
MW: 134.15 g/mol
Boiling Pt: 122…126 °C
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD08062432
CAS Number: 55816-69-8
UN: 1992
ADR: 3+6.1 III