(3-Methoxy-5-(trifluoromethyl)phenyl)acetic acid 98%
Supplier: Apollo Scientific
Warning
PC49342-1G PC49342-5G PC49342-10G
APOSPC49342-1GEA 203 EUR
APOSPC49342-1G APOSPC49342-5G APOSPC49342-10G
(3-Methoxy-5-(trifluoromethyl)phenyl)acetic acid 98%
(3-Methoxy-5-(trifluoromethyl)phenyl)acetic acid
Formula: C₁₀H₉F₃O₃
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD09025416
CAS Number: 916421-04-0