4-(Trifluoromethyl)piperidine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Trifluoromethylpiperidine
PC49502-5G PC49502-1G PC49502-25G
APOSPC49502-5GEA 181 EUR
APOSPC49502-5G APOSPC49502-1G APOSPC49502-25G
4-(Trifluoromethyl)piperidine 97%
4-(Trifluoromethyl)piperidine
Formula: C₆H₁₀F₃N
MW: 153.15 g/mol
Boiling Pt: 135 °C
Density: 1.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00102068
CAS Number: 657-36-3
UN: 1993
ADR: 3 III