Methyl-2-fluoroacrylate 97%
Supplier: Apollo Scientific
Danger
PC4967-25G PC4967-5G
APOSPC4967-25GEA 147 EUR
APOSPC4967-25G APOSPC4967-5G
Methyl-2-fluoroacrylate 97%
Methyl-2-fluoroacrylate
Formula: C₄H₅FO₂
Boiling Pt: 95 °C/760mmHg
Melting Pt: –42 °C
Density: 1.114 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039286
CAS Number: 2343-89-7
UN: 3286
ADR: 3+6.1+8,II