3-Bromo-5-(trifluoromethoxy)phenol 97%
Supplier: Apollo Scientific
Warning
PC499005-1G PC499005-250MG PC499005-5G
APOSPC499005-1GEA 162 EUR
APOSPC499005-1G APOSPC499005-250MG APOSPC499005-5G
3-Bromo-5-(trifluoromethoxy)phenol 97%
3-Bromo-5-(trifluoromethoxy)phenol
Formula: C₇H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD15142929
CAS Number: 1197239-47-6