2-Methoxy-4-(trifluoromethyl)nicotinic acid 95%
Supplier: Apollo Scientific
Warning
PC50022-1G PC50022-250MG
APOSPC50022-1GEA 671 EUR
APOSPC50022-1G APOSPC50022-250MG
2-Methoxy-4-(trifluoromethyl)nicotinic acid 95%
2-Methoxy-4-(trifluoromethyl)nicotinic acid
Formula: C₈H₆F₃NO₃
Melting Pt: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD14584806
CAS Number: 1221792-53-5