2-Fluoro-3-nitroaniline
Supplier: Apollo Scientific
|
Warning
|
PC501674-1G
PC501674-250MG
PC501674-5G
APOSPC501674-1GEA
114
EUR
APOSPC501674-1G
APOSPC501674-250MG
APOSPC501674-5G
2-Fluoro-3-nitroaniline
2-Fluoro-3-nitroaniline
Formula:
C₆H₅FN₂O₂ Melting Pt: 114…116 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11046757 CAS Number: 21397-11-5 |