Methyl-2-bromo-5-(trifluoromethoxy)benzoate 95%
Supplier: Apollo Scientific
PC502712-1G PC502712-5G
APOSPC502712-1GEA 74.1 EUR
APOSPC502712-1G APOSPC502712-5G
Methyl-2-bromo-5-(trifluoromethoxy)benzoate 95%
Methyl 2-bromo-5-(trifluoromethoxy)benzoate
Formula: C₉H₆BrF₃O₃
MW: 299.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025999
CAS Number: 1150114-81-0