2-Methoxy-5-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Warning
PC5063X-1G PC5063X-5G PC5063X-10G
APOSPC5063X-1GEA 14.7 EUR
APOSPC5063X-1G APOSPC5063X-5G APOSPC5063X-10G
2-Methoxy-5-(trifluoromethyl)pyridine 98%
2-Methoxy-5-(trifluoromethyl)pyridine
Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153203
CAS Number: 175277-45-9