2,3-Dichloro-6-fluorobenzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC51143-25G PC51143-5G PC51143-1G
APOSPC51143-25GEA 386 EUR
APOSPC51143-25G APOSPC51143-5G APOSPC51143-1G
2,3-Dichloro-6-fluorobenzaldehyde 97%
2,3-Dichloro-6-fluorobenzaldehyde
Formula: C₇H₃Cl₂FO
MW: 193 g/mol
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD04115870
CAS Number: 95399-95-4