1,1,5,5-Tetrafluoro-2,4-pentanedione
Supplier: Apollo Scientific
Warning
PC51157-25G PC51157-5G
APOSPC51157-25GEA 256 EUR
APOSPC51157-25G APOSPC51157-5G
1,1,5,5-Tetrafluoro-2,4-pentanedione
1,1,5,5-Tetrafluoro-2,4-pentanedione
Formula: C₅H₄F₄O₂
MW: 172.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419795
CAS Number: 70086-62-3
UN: 1993
ADR: 3 III