Methyl-2-(trifluoromethyl)benzoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-(trifluoromethyl)benzoate
PC5171Y-25G PC5171Y-5G PC5171Y-1G
APOSPC5171Y-25GEA 130 EUR
APOSPC5171Y-25G APOSPC5171Y-5G APOSPC5171Y-1G
Methyl-2-(trifluoromethyl)benzoate 97%
Methyl 2-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
Boiling Pt: 98…100 °C
Density: 1.306 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236050
CAS Number: 344-96-7