Methyl-3-(trifluoromethyl)benzoate 95%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-(trifluoromethyl)benzoate
PC5171Z-100G PC5171Z-25G PC5171Z-500G
APOSPC5171Z-100GEA 46.4 EUR
APOSPC5171Z-100G APOSPC5171Z-25G APOSPC5171Z-500G
Methyl-3-(trifluoromethyl)benzoate 95%
Methyl 3-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
Boiling Pt: 93…95 °C/20mmHg
Density: 1.295 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00043543
CAS Number: 2557-13-3
UN: 3272
ADR: 3,III