Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Trifluoromethyl)-3,3,3-trifluoropropionic Acid Methyl Ester, Methyl 2,2-Bis(trifluoromethyl)acetate, 2,2-Bis(trifluoromethyl)acetic Acid Methyl Ester
PC5184E-5G PC5184E-1G PC5184E-25G
APOSPC5184E-5GEA 190 EUR
APOSPC5184E-5G APOSPC5184E-1G APOSPC5184E-25G
Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate 97%
Methyl-2-(trifluoromethyl)-3,3,3-trifluoropropionate
Formula: C₅H₄F₆O₂
MW: 210.08 g/mol
Boiling Pt: 90…91 °C
Density: 1.604 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00054663
CAS Number: 360-54-3
UN: 3272
ADR: 3 III