2-Chloro-5-chlorosulphonyl-4-fluorobenzoic acid 98%
Supplier: Apollo Scientific
PC520514-100G PC520514-50G
APOSPC520514-100GEA 757 EUR
APOSPC520514-100G APOSPC520514-50G
2-Chloro-5-chlorosulphonyl-4-fluorobenzoic acid 98%
2-Chloro-5-chlorosulphonyl-4-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₄S
MW: 273.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321365
CAS Number: 264927-50-6