2,3-Dimethyl-4-fluoroaniline 97%
Supplier: Apollo Scientific
Synonyms: 4-Fluoro-2,3-dimethylaniline, 4-Fluoro-2,3-xylidine, 3-Amino-6-fluoro-o-xylene
PC520532-1G PC520532-5G
APOSPC520532-1GEA 243 EUR
APOSPC520532-1G APOSPC520532-5G
2,3-Dimethyl-4-fluoroaniline 97%
2,3-Dimethyl-4-fluoroaniline
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972870
CAS Number: 1737-68-4