2-(Trifluoromethyl)-1H-indole 97%
Supplier: Apollo Scientific
Warning
PC520534-1G PC520534-250MG
APOSPC520534-1GEA 168 EUR
APOSPC520534-1G APOSPC520534-250MG
2-(Trifluoromethyl)-1H-indole 97%
2-(Trifluoromethyl)-1H-indole
Formula: C₉H₆F₃N
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD04972104
CAS Number: 51310-54-4