2,4-Difluororesorcinol 97%
Supplier: Apollo Scientific
Synonyms: 2,4-Difluorobenzene-1,3-diol
PC520578-1G PC520578-500MG PC520578-5G
APOSPC520578-1GEA 203 EUR
APOSPC520578-1G APOSPC520578-500MG APOSPC520578-5G
2,4-Difluororesorcinol 97%
2,4-Difluororesorcinol
Formula: C₆H₄F₂O₂
Storage Temperature: Freezer
CAS Number: 195136-71-1
UN: 2811
ADR: 6.1,III