2,2'-Bis(trifluoromethyl)biphenyl 95%
Supplier: Apollo Scientific
Warning
PC520582-10G PC520582-5G
APOSPC520582-10GEA 1230 EUR
APOSPC520582-10G APOSPC520582-5G
2,2'-Bis(trifluoromethyl)biphenyl 95%
2,2'-Bis(trifluoromethyl)biphenyl
Formula: C₁₄H₈F₆
MW: 290.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836618
CAS Number: 567-15-7