(S)-2-(Trifluoromethyl)piperidine 97%
Supplier: Apollo Scientific
PC520605-1G PC520605-5G
APOSPC520605-1GEA 118 EUR
APOSPC520605-1G APOSPC520605-5G
(S)-2-(Trifluoromethyl)piperidine 97%
(S)-2-(Trifluoromethyl)piperidine
Formula: C₆H₁₀F₃N
Storage Temperature: Ambient
MDL Number: MFCD11226597
CAS Number: 154727-51-2