(4,4-Difluorocyclohexyl)methanol 97%
Supplier: Apollo Scientific
Warning
PC520664-25G PC520664-5G
APOSPC520664-25GEA 277 EUR
APOSPC520664-25G APOSPC520664-5G
(4,4-Difluorocyclohexyl)methanol 97%
(4,4-Difluorocyclohexyl)methanol
Formula: C₇H₁₂F₂O
Boiling Pt: 207…213 °C
Density: 1.1543 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD11847775
CAS Number: 178312-48-6