4-(1,1-Difluoroethyl)pyridine 95%
Supplier: Apollo Scientific
PC520687-1G PC520687-25G PC520687-5G
APOSPC520687-1GEA 199 EUR
APOSPC520687-1G APOSPC520687-25G APOSPC520687-5G
4-(1,1-Difluoroethyl)pyridine 95%
4-(1,1-Difluoroethyl)pyridine
Formula: C₇H₇F₂N
Storage Temperature: Ambient
MDL Number: MFCD12404950
CAS Number: 114490-29-8