2-Bromo-1-(difluoromethyl)-4-fluorobenzene 98%
Supplier: Apollo Scientific
PC520901-25G PC520901-5G
APOSPC520901-25GEA 734 EUR
APOSPC520901-25G APOSPC520901-5G
2-Bromo-1-(difluoromethyl)-4-fluorobenzene 98%
2-Bromo-1-(difluoromethyl)-4-fluorobenzene
Formula: C₇H₄BrF₃
Storage Temperature: Ambient
MDL Number: MFCD06657961
CAS Number: 845866-81-1