2-Bromo-4-(1,1-difluoroethyl)-1-fluorobenzene 97%
Supplier: Apollo Scientific
PC520910-1G PC520910-5G
APOSPC520910-1GEA 144 EUR
APOSPC520910-1G APOSPC520910-5G
2-Bromo-4-(1,1-difluoroethyl)-1-fluorobenzene 97%
2-Bromo-4-(1,1-difluoroethyl)-1-fluorobenzene
Formula: C₈H₆BrF₃
Storage Temperature: Ambient
MDL Number: MFCD11845795
CAS Number: 1138444-85-5