2-Amino-3-(trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Warning
PC5257-1G PC5257-5G PC5257-10G
APOSPC5257-1GEA 21.9 EUR
APOSPC5257-1G APOSPC5257-5G APOSPC5257-10G
2-Amino-3-(trifluoromethoxy)benzoic acid 97%
2-Amino-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₆F₃NO₃
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD08669644
CAS Number: 561304-41-4