1-Iodoperfluorbutane 98%
Supplier: Apollo Scientific
Warning
Synonyms: Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane
PC5310-100G PC5310-25G PC5310-500G
APOSPC5310-100GEA 120 EUR
APOSPC5310-100G APOSPC5310-25G APOSPC5310-500G
1-Iodoperfluorbutane 98%
1-Iodoperfluorbutane
Formula: C₄F₉I
MW: 345.93 g/mol
Boiling Pt: 64…66 °C
Melting Pt: -2
Density: 2.01 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001062
CAS Number: 423-39-2
EINECS: 207-025-8