2-(Trifluoromethyl)-1,6-naphthyridine-3-carboxylic acid 95+%
Supplier: Apollo Scientific
Warning
PC5321-1G PC5321-5G PC5321-250MG
APOSPC5321-1GEA 331 EUR
APOSPC5321-1G APOSPC5321-5G APOSPC5321-250MG
2-(Trifluoromethyl)-1,6-naphthyridine-3-carboxylic acid 95+%
2-(Trifluoromethyl)-1,6-naphthyridine-3-carboxylic acid
Formula: C₁₀H₅F₃N₂O₂
MW: 242.16 g/mol
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00202962
CAS Number: 240408-97-3