3-Cyano-4-fluorobenzenesulphonamide 97%

Supplier: Apollo Scientific

PC53374-10G PC53374-1G PC53374-5G
APOSPC53374-10GEA 793 EUR
APOSPC53374-10G APOSPC53374-1G APOSPC53374-5G
3-Cyano-4-fluorobenzenesulphonamide 97%
3-Cyano-4-fluorobenzenesulphonamide
Formula: C₇H₅FN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD16705414
CAS Number: 928139-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR