1-Fluorooctane 98%
Supplier: Apollo Scientific
Warning
Synonyms: Octyl fluoride
PC5360-25G PC5360-5G
APOSPC5360-25GEA 725 EUR
APOSPC5360-25G APOSPC5360-5G
1-Fluorooctane 98%
1-Fluorooctane
Formula: C₈H₁₇F
Boiling Pt: 142…146 °C
Density: 0.814 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00013582
CAS Number: 463-11-6
EINECS: 207-333-2
UN: 1993
ADR: 3,III