5-Chloro-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile
Supplier: Apollo Scientific
PC5499-1G PC5499-500MG
APOSPC5499-1GEA 137 EUR
APOSPC5499-1G APOSPC5499-500MG
5-Chloro-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile
5-Chloro-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile
Formula: C₈H₄ClF₃N₂O
MW: 236.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11501054
CAS Number: 1092352-68-5