2,4-Difluoropyridine 98%
Supplier: Apollo Scientific
Warning
PC5506-1G PC5506-25G PC5506-5G
APOSPC5506-1GEA 21.5 EUR
APOSPC5506-1G APOSPC5506-25G APOSPC5506-5G
2,4-Difluoropyridine 98%
2,4-Difluoropyridine
Formula: C₅H₃F₂N
Boiling Pt: 107 °C/760mmHg
Density: 1.26 g/cm³ (25 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD04114162
CAS Number: 34941-90-7
UN: 1993
ADR: 3,III