Pentafluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3,4,5,6-Pentafluorobenzonitrile
PC5510-100G PC5510-500G PC5510-5G
APOSPC5510-100GEA 79.9 EUR
APOSPC5510-100G APOSPC5510-500G APOSPC5510-5G
Pentafluorobenzonitrile 98%
Pentafluorobenzonitrile
Formula: C₇F₅N
MW: 193.08 g/mol
Boiling Pt: 161…164 °C
Melting Pt: 44256 °C
Density: 1.53 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00001775
CAS Number: 773-82-0
EINECS: 212-259-9
UN: 1993
ADR: 3 III