Methyl-3-methoxy-2-(trifluoromethyl)propionate 97%
Supplier: Apollo Scientific
Warning
PC5511-1G PC5511-5G PC5511-250MG
APOSPC5511-1GEA 323 EUR
APOSPC5511-1G APOSPC5511-5G APOSPC5511-250MG
Methyl-3-methoxy-2-(trifluoromethyl)propionate 97%
Methyl-3-methoxy-2-(trifluoromethyl)propionate
Formula: C₆H₉F₃O₃
MW: 186.13 g/mol
Melting Pt: 242 °C
Storage Temperature: Ambient
MDL Number: MFCD04038322
CAS Number: 359-88-6
UN: 3272
ADR: 3 III