(E)-1-Methoxy-3,3,3-trifluoropropene 97%
Supplier: Apollo Scientific
Warning
PC5592-1G PC5592-5G
APOSPC5592-1GEA 199 EUR
APOSPC5592-1G APOSPC5592-5G
(E)-1-Methoxy-3,3,3-trifluoropropene 97%
(E)-1-Methoxy-3,3,3-trifluoropropene
Formula: C₄H₅F₃O
Boiling Pt: 61…63 °C
Density: 1.1568 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD04038316
CAS Number: 26885-71-2
UN: 1993
ADR: 3,III