2,3-Difluorobenzhydrazide 98%
Supplier: Apollo Scientific
Warning
PC5597-1G PC5597-5G PC5597-25G
APOSPC5597-1GEA 50.9 EUR
APOSPC5597-1G APOSPC5597-5G APOSPC5597-25G
2,3-Difluorobenzhydrazide 98%
2,3-Difluorobenzhydrazide
Formula: C₇H₆F₂N₂O
MW: 172.14 g/mol
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD02183536
CAS Number: 387358-53-4