Potassium-4-(trifluoroboratomethyl)thiomorpholine
Supplier: Apollo Scientific
Warning
Synonyms: Potassium trifluoro(thiomorpholinomethyl)borate
PC56184-1G PC56184-5G
APOSPC56184-1GEA 181 EUR
APOSPC56184-1G APOSPC56184-5G
Potassium-4-(trifluoroboratomethyl)thiomorpholine
Potassium 4-(trifluoroboratomethyl)thiomorpholine
Formula: C₅H₁₀BF₃KNS
Storage Temperature: Freezer
MDL Number: MFCD10700162
CAS Number: 1150654-80-0