1-(Difluoromethyl)-4-nitro-benzene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Nitrobenzal fluoride
PC57047-1G PC57047-5G
APOSPC57047-1GEA 72.5 EUR
APOSPC57047-1G APOSPC57047-5G
1-(Difluoromethyl)-4-nitro-benzene
1-(Difluoromethyl)-4-nitrobenzene
Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Boiling Pt: 80…84 °C/1.5mmHg
Melting Pt: 21…26 °C
Density: 1.39 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD16659620
CAS Number: 29848-57-5