2-Methoxy-5-(trifluoromethyl)benzoic acid 98%
Supplier: Apollo Scientific
Warning
PC5709-25G PC5709-5G PC5709-10G
APOSPC5709-25GEA 166 EUR
APOSPC5709-25G APOSPC5709-5G APOSPC5709-10G
2-Methoxy-5-(trifluoromethyl)benzoic acid 98%
2-Methoxy-5-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Melting Pt: 107…111 °C
Storage Temperature: Ambient
MDL Number: MFCD01862091
CAS Number: 4864-01-1