2-Bromo-2'-(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[2-(trifluoromethyl)phenyl]-1-ethanone
PC5799-10G PC5799-1G PC5799-5G
APOSPC5799-10GEA 328 EUR
APOSPC5799-10G APOSPC5799-1G APOSPC5799-5G
2-Bromo-2'-(trifluoromethyl)acetophenone
2-Bromo-2'-(trifluoromethyl)acetophenone
Formula: C₉H₆BrF₃O
MW: 267.04 g/mol
Boiling Pt: 52 °C/0.03mmHg
Melting Pt: 24…32 °C
Storage Temperature: Ambient
MDL Number: MFCD03094304
CAS Number: 54109-16-9
UN: 3261
ADR: 8 III